Imprimer
Hotplate, RC 5
Fournisseur: VELP SCIENTIFIC
F20700610
VELPF20700610EA 237 EUR
VELPF20700610
Hotplate, RC 5
Appareils chauffants Plaques chauffantes
Single-position hotplate with resistant aluminum top plate.
  • Excellent heating performance up to 310 °C with homogeneous temperature
  • The internal electronic parts are protected by the insulating disk which resists even the most challenging environments and applications (IP 42 protection)
  • Safety functions to protect users in daily operations, also with analogue models
The RC 5 is equipped with a 135 mm diameter aluminum alloy top plate ensuring a great temperature homogeneity and optimum heat transfer across the entire surface.
It is reliable forMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR