Imprimer
Ceramic Magnetic Hotplate Stirrer, AREC 7
Fournisseur: VELP SCIENTIFIC
F20500680
VELPF20500680EA 440 EUR
VELPF20500680
Ceramic Magnetic Hotplate Stirrer, AREC 7
Agitateurs Agitateurs magnétiques chauffants
Analogue hotplate stirrer, equipped with a 177,8 mm square ceramic plate, safe and reliable, designed to deliver excellent heating and stirring performance for basic laboratory applications.
  • Advanced heating technology ensures temperature homogeneity and stability from room temperature up to 550 °C
  • Powerful brushless ensuring power and resistance over time
  • Stirs up to 15 L volume (H₂O) from 100 up to 1500 min⁻¹
  • SpeedServo™ counter reaction technology maintains the speed constant as viscosity changes
  • The robust 177,8 mm ceramic top offers exceptional
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR