Imprimer
Analytical HPLC columns, Microsorb™ Cyano
Fournisseur: VARIAN
R0089800C5 R0089800D5 R0086800D5 R0086800C8 R0086800E3 R0086800C5
VARIR0089800C5EA 963 EUR
VARIR0089800C5 VARIR0089800D5 VARIR0086800D5 VARIR0086800C8 VARIR0086800E3 VARIR0086800C5
Analytical HPLC columns, Microsorb™ Cyano
Colonnes pour chromatographie
Microsorb™ Cyano, a cyanopropylsilane-covered silica, is unique in that it spans both the normal and reversed phase separation modes. When used with aqueous organic mobile phases, compounds elute in an order typical of reversed phase, with polar compounds eluting quickly and hydrophobic compounds being more retained.
This retention makes Microsorb™ Cyano columns very useful for separating compounds that are too hydrophobic for C18, and that do not dissolve well in organic solvents. Cyano columns can also be used with typical normal phase solvents to separate compounds on the basis of polar interactions.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR