Imprimer
Chemistry roots pump units
Fournisseur: VACUUBRAND
691035 691036 691037 691040 691041 691042 691050 691051 691052 691060 691061 691062
VACU691035EA 0 EUR
VACU691035 VACU691036 VACU691037 VACU691040 VACU691041 VACU691042 VACU691050 VACU691051 VACU691052 VACU691060 VACU691061 VACU691062
Chemistry roots pump units
Pompe Pompes à vide à membrane
Pump units with the combination of an oil-free chemistry diaphragm pump with a Roots pump leads to a significant increase in pumping speed and improves ultimate vacuum by an order of magnitude.
  • Low ultimate vacuum <0,2 mbar
  • Corrosion resistance and active pollution control
  • Virtually oil-free vacuum compared to rotary vane pumps
  • Measurably reduced backflow of oil due to oil-free pump chamber of roots pump
  • Very high pumping speed from 12 to 40 m³/h, maximum between 0,2 and 100 mbar
The dry and oil-free compression and their good pumping speed,More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR