Imprimer
Welding spectacles, 9104
Fournisseur: UVEX
9104.041
UVEX9104.041EA 57.2 EUR
UVEX9104.041
Welding spectacles, 9104
Lunettes
Compact flip-up welding safety spectacles with a close fit and anatomically shaped nose bridge, side shields and integrated brow guard are designed for increased protection with flip-up and easily replaceable lens.
  • Perfect colour recognition thanks to uvex sunglare filters
  • Reliable UV 400 and IR protection (flip-up lens)
  • Minimum damage from welding sparks
  • Side protection and integrated brow guard frame design
  • Fully adjustable side arms
  • Adjustable, soft and comfortable ear pieces
The specially moulded frame edge provides protection against particlesMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR