Imprimer
Outlet valve
Fournisseur: USBECK Laborgeräte
69500
USBE69500EA 23.5 EUR
USBE69500
Outlet valve
Vannes Vannes de régulation d'air
Outlet valve with M10×1 outside thread.
Certifications: According to DIN EN 417:2012 and CE-0085CQ0255
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR