Imprimer
Cerex® THC SPE Columns
Fournisseur: Tecan
682-0101R 682-0353C 682-0503C 682-0506C
TCAT682-0101REA 0 EUR
TCAT682-0101R TCAT682-0353C TCAT682-0503C TCAT682-0506C
Cerex® THC SPE Columns
Colonnes pour chromatographie
Polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation. Specially formulated for THC/THCA testing.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR