Imprimer
SPE Column, NBE OFX
Fournisseur: Tecan
633-0031R-NBE
TCAT633-0031R-NBEEA 0 EUR
TCAT633-0031R-NBE
SPE Column, NBE OFX
Colonnes pour chromatographie
Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.
Plus About VWR