Imprimer
SPE Columns, NBE HPSCX
Fournisseur: Tecan
427-0021R-NBE 427-0051R-NBE
TCAT427-0021R-NBEEA 0 EUR
TCAT427-0021R-NBE TCAT427-0051R-NBE
SPE Columns, NBE HPSCX
Colonnes pour chromatographie
Mixed mode sorbent with both reverse-phase and strong cationic exchange chemistries.
  • Ion-exchange
  • Liquid handler compatible
Its high surface area provides excellent capacity and a high selectivity for basic analytes. Ideal for drug monitoring, screening, identification, confirmation and quantitation.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR