Imprimer
EasySep™ Human Naïve B Cell Enrichment Kits
Fournisseur: STEMCELL Technologies
19254 19254RF
STMC19254EA 1138 EUR
STMC19254 STMC19254RF
EasySep™ Human Naïve B Cell Enrichment Kits
Milieux de séparation cellulaire
Immunomagnetic isolation of untouched human naïve B cells
  • Fast, easy-to-use and column-free
  • Up to 98% purity
  • Isolated cells are untouched
The EasySep™ Human Naïve B Cell Enrichment Kit is designed to isolate naïve B cells from peripheral blood mononuclear cells by negative selection. Unwanted cells are targeted for removal with Tetrameric Antibody Complexes recognising non-naïve B cells and dextran-coated magnetic particles. The labelled cells are separated using the EasySep™ magnet without the use of columns. The desired cells are poured off into aMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR