Imprimer
StemSpan™ CC110 Culture Supplement
Fournisseur: STEMCELL Technologies
02697
STMC02697EA 965 EUR
STMC02697
StemSpan™ CC110 Culture Supplement
Milieux de culture cellulaire
Serum-free culture supplement for expansion of human hematopoietic cells.
StemSpan™ CC110 contains a combination of early-acting recombinant human (rh) cytokines formulated to support the proliferation of human hematopoietic cells. It is supplied as a 100X concentrate. When added to serum-free medium, StemSpan™ CC110 promotes the expansion of CD34+ cells isolated from human cord blood and bone marrow. StemSpan™ CC110 stimulates similar expansion of CD34+ cells compared to StemSpan™ CC100, but with higher purity and is meant for use in short-term cultures to activate stem cell and immature progenitor cycling, without necessarily promoting the proliferation and differentiation of later progenitors.
We recommend using StemSpan™ CC110 in combination with any of the following StemSpan™ media:
StemSpan™ SFEM
StemSpan™ SFEM II
StemSpan™-XF
StemSpan™-AOF
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR