Imprimer
NGL-IMPACT® CH1 Affinity Ligand ELISA Kit
Fournisseur: Spectrum Laboratories
9-EL-0040
SPEC9-EL-0040EA 1450 EUR
SPEC9-EL-0040
NGL-IMPACT® CH1 Affinity Ligand ELISA Kit
Tests Tests ELISA
The NGL-IMPACT® CH1 Affinity Ligand ELISA Kit provides accurate and precise quantitation for downstream processes that utilise a Purolite Praesto Jetted 70 CH1 capture step.
  • Exact matched reference standard (NGL-IMPACT® CH1 ligand)
  • Recovery: 80 to 120%
  • Accuracy and precision: inter-assay ±15%, intra-assay ±15%
Kit contents: Anti-CH1 antibody coated 96 well microtiter plate, Reagent A: sample diluent (5X), Reagent B: NGL-IMPACT® CH1 Ligand standard solution, Reagent C: rabbit anti-CH1: biotin probe, Reagent D: streptavidin: peroxidase conjugate, Reagent E: TMB peroxidase substrate, PBS packs (quantity 2)
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR