Imprimer

Citric acid, anhydre ≥99.5%, granuleux FCC

Fournisseur: Spectrum Chemical

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid

C1281-500GM
SPCMC1281-500GMEA 179 EUR
SPCMC1281-500GM
Citric acid, anhydre ≥99.5%, granuleux FCC
Acide citrique

Citric Acid, Anhydrous, Granular, FCC is the most widely used organic acid and pH-control agent in foods and beverages. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formule: HOC(COOH)(CH₂COOH)₂
MW: 192.13 g/mol
Point d'ébullition: 310
Température de stockage: Température ambiante
Numéro MDL: MFCD00011669
Numéro CAS: 77-92-9
Merck Index: 13,02350
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d60f5a9b8e6c6dfb7eda04e1a1d5d6d971f92444&vid=caf4f9effc7fd22b942f7f92be770b06e40637a4&prid=web&clid=SAPDAM

Commander maintenant

Résultats des tests de spécification

Assay (C₆H₈O₇ ;Anhyd.,Basis) 99.5 - 100.5%
Lead (Pb) max. 0.5 mg/kg
Oxalate (C₂O₄) To Pass Test
Readily Carbonizable Substances To Pass Test
Residue On Ignition max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR