Imprimer
LYNX 4000 and 6000 Superspeed Centrifuges with GreenCool Technology LYNX 4000 and 6000
Fournisseur: SORVALL
75008580 75008581 75008591 75008592
SORV75008580EA 44940 EUR
SORV75008580 SORV75008581 SORV75008591 SORV75008592
LYNX 4000 and 6000 Superspeed Centrifuges with GreenCool Technology LYNX 4000 and 6000
Centrifugeuses Centrifugeuses sur pieds
Thermo Scientific™ LYNX 4000 and 6000 Superspeed Centrifuges with GreenCool technology bring enhanced sustainability to research and bioprocessing laboratories. They are equipped with a natural refrigerant (CO₂) cooling system, reducing environmental impact without compromising performance. Their lighter weight and reduced noise levels help improve the laboratory environment.
  • Global Warming Potential (GWP) of 1, up to 13% lower energy consumption
  • Up to 4 L capacity and RCF up to 100,605 ×g
  • Auto-Lock rotor exchange, secure rotor exchange in less than 3
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR