Imprimer
TriLink® 2',3'-Dideoxythymidine-5'-O-(1-Thiotriphosphate)
Fournisseur: TriLink BioTechnologies
N-8012-1 N-8012-10 N-8012-5
SOLUN-8012-1EA 94.3 EUR
SOLUN-8012-1 SOLUN-8012-10 SOLUN-8012-5
TriLink® 2',3'-Dideoxythymidine-5'-O-(1-Thiotriphosphate)
Réactifs pour acides nucléiques Nucléotides
2',3'-Dideoxythymidine-5'-O-(1-Thiotriphosphate) is a sugar modified nucleoside triphosphate where the 2' and 3' hydroxyl groups are absent, resulting in chain termination. This ddTTP analog also contains a 1-Thiotriphosphate which results in a nuclease resistant phosphorothioate linkage. The inability of polymerases to extend from a dideoxy nucleotide causes chain termination and is useful in antiviral research and in a variety of biotechnology applications.
  • Grade: RUO
Attention: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d254b8bd584b1c7b77313ac7837979e4753596ad&vid=d254b8bd584b1c7b77313ac7837979e4753596ad&prid=web&clid=SAPDAM
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR