Imprimer
Sterilin™ Sample Bags, Homogeniser
Fournisseur: STERILIN
S400
SLINS400EA 170 EUR
SLINS400
Sterilin™ Sample Bags, Homogeniser
Sachets Sachets d'échantillonnage
Made of food-grade heavy-gauge polyethylene, these bags are double sealed for strength and reliability ensuring safe homogenisation of samples and are ideal for homogenising food samples prior to microbiological analysis. They are manufactured from food grade low density PE and are gamma irradiated to ensure the sterility.
  • Suitable for use in all leading homogeniser machines
  • Heavy gauge polyethylene for strength
  • Double-sealed for strength and reliability
  • Ensuring safe processing of samples
  • Manufactured from food-grade heavy-guage polyethylene for
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR