Imprimer
Film applicators, multi-functional
Fournisseur: Industrial Physics
VF1550
SIMEVF1550EA 631.02 EUR
SIMEVF1550
Film applicators, multi-functional
Applicateurs de films Applicateurs de film pour peintures
Multi-functional film applicator (Octoplex) with 8 pre-defined thickness sides for applying paint films of 25, 50, 75, 100, 125, 150, 175, 200 µm. Film width 60 mm. The high-grade stainless steel will not be affected by acid or base elements.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR