Imprimer
Film applicators, spiral type
Fournisseur: Industrial Physics
VF0114 VF0120 VF0119 VF0118 VF0117 VF0115 VF0113 VF0111 VF0110 VF0109 VF0108 VF0107 VF0106 VF0105 VF0104 VF0103 VF0102 VF0101 VF0116 VF0100
SIMEVF0114EA 164.46 EUR
SIMEVF0114 SIMEVF0120 SIMEVF0119 SIMEVF0118 SIMEVF0117 SIMEVF0115 SIMEVF0113 SIMEVF0111 SIMEVF0110 SIMEVF0109 SIMEVF0108 SIMEVF0107 SIMEVF0106 SIMEVF0105 SIMEVF0104 SIMEVF0103 SIMEVF0102 SIMEVF0101 SIMEVF0116 SIMEVF0100
Film applicators, spiral type
Applicateurs de films Applicateurs de film pour peintures
Spiral type applicator with a film width of 250 mm and available in ranges from 4 to 200 μm. Ideal for application of film onto thin materials, such as sheets or plastic. Also works on flexible substrates, and with motorised film applicators. The high grade stainless steel will not be affected by acid and base elements.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR