Imprimer
Surface probes, clamp-type, for Curvex datalogger
Fournisseur: Industrial Physics
CX2040 CX2041 CX2030 CX2045
SIMECX2040EA 201.79 EUR
SIMECX2040 SIMECX2041 SIMECX2030 SIMECX2045
Surface probes, clamp-type, for Curvex datalogger
Sondes de température
Small and elegant clamp-type probes to measure temperature of any object.
  • Max. object thickness is 20 mm
  • Energy and/or water efficient
  • Recycled/renewable content
Silver tipped sensor is thermally isolated from the clamp by ceramics. The capton-wrapped, metal braid and PTFE coated probe cables last significantly longer than other designs.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR