Imprimer
Film applicators, automatic, standard
Fournisseur: Industrial Physics
AB4120 AB4220 AB4320 AB4400 AB4420
SIMEAB4120EA 7290 EUR
SIMEAB4120 SIMEAB4220 SIMEAB4320 SIMEAB4400 SIMEAB4420
Film applicators, automatic, standard
Applicateurs de films Applicateurs de film pour peintures
The Automatic Film Applicator Standard provides a reliable basis to apply coating films to test charts, panels or foils in a uniform and reproducible way in order to eliminate variations caused by human factors.
  • Paper clamp/vacuum manually operable
  • Full colour display
  • Triple i control (intelligent illumination interface) and easy to update software
  • Drying time recorder option
  • Charts up to A3 size
  • Preset range option
  • Compatible with a large range of application tools
Reproducible
Variations in speed, pressure and direction of draw down causeMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR