Imprimer
Portable hygiene monitoring system, MVP ICON®
Fournisseur: Merck
64003-100 78087-00 78088-00 78089-00
SIAL64003-100EA 408 EUR
SIAL64003-100 SIAL78087-00 SIAL78088-00 SIAL78089-00
Portable hygiene monitoring system, MVP ICON®
Systèmes de surveillance Tests de contrôle de l'hygiène
MVP ICON® portable hygiene monitoring system – setting a new standard.
  • Multi-variable platform – allows for monitoring, tracking and management of ATP as indicator of hygiene of the surfaces and other HACCP parameters like conductivity, sanitiser concentration, pH and temperature
  • Quick and easy to use – The MVP ICON® features a touch screen display with full colour graphics, making it highly visible and easy to use, even while wearing latex gloves
  • On-site calibration – for all parameters, including ATP, to ensure on-going accuracy of measurements and
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR