Imprimer
Spray Bottles, PP
Fournisseur: SEMADENI
12050
SEMA12050EA 0 EUR
SEMA12050
Spray Bottles, PP
Flacons Flacons pulvérisateurs
Made from PP.
  • White
  • Reusable
Sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle, which can be regulated from the finest mist (nebulising) to a precise liquid jet.
Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR