Imprimer
Segmented polariser set for VisiLED ring light, S80-55
Fournisseur: SCHOTT AG Lighting and imaging
400555
SCOC400555EA 699 EUR
SCOC400555
Segmented polariser set for VisiLED ring light, S80-55
Éclairages Éclairages annulaires
The segmented polariser set has the ability to switch between polarised and non-polarised light, without any mechanical movements. The set is only compatible to VisiLED ringlight S80 55, together with the MC 1500 or the MC 1100 controller.
  • Very helpful for all applications that need polarised and non-polarised light, such as cleanliness inspections
  • Software control possible (MC 1500)
  • Intensities independently adjustable for polarisation and brightfield (MC 1500)
Designed for use with VisiLED ring light S80-55, together with the MC 1500 or the MC 1100More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR