Imprimer
Biochrom Biowave 3 with Printer
Fournisseur: BIOCHROM
80-3007-33
SCLI80-3007-33EA 7720 EUR
SCLI80-3007-33
Biochrom Biowave 3 with Printer
Spectrophotomètres et Photomètres Spectrophotomètres UV-Visible
The Biowave 3 series offers all the benefits and capabilities of a research grade Life Science spectrophotometer with robust design and reasonable cost that also makes it perfect for students.
  • Split beam (RBC) UV/Vis spectrophotometer 190 to 1100 nm (3/5 nm bandwidth)
  • Pre-programmed methods for DNA, RNA, oligos, proteins and cell density
  • Nucleic acid scans for purity checking
  • Low volume capability with microvolume cuvettes
  • Novel optics with a xenon source for long lamp lifetime and low running costs
  • Lightweight and stackable for storage
Results may beMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR