Imprimer
Cleaning cloth, low-lint, Tork Industrial
Fournisseur: Essity
190578
SCAA190578EA 189 EUR
SCAA190578
Cleaning cloth, low-lint, Tork Industrial
Lingettes Lingettes et essuie-mains à usage unique Lingettes en papier pour tâches délicates
This extremely low lint cloth with anti-static properties is ideal for use with solvents when cleaning smooth surfaces. Made from 100% PP.
  • Perfect for working with sensitive surfaces due to extreme the low-lint and antistatic properties
  • Strong texture for excellent oil and water absorption
  • Tork Easy Handling™ for easier carrying, opening and disposing of packaging
  • The silicone-free cloth ensures that there are no residues on a surface
  • Food contact approved
Suitable for use in clean condition areas, printing industry, precision engineering, electronicsMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR