Imprimer
X-ray and metal detectable ladles
Fournisseur: Sampling Systems
M1033B-50
SAMPM1033B-50EA 20.2 EUR
SAMPM1033B-50
X-ray and metal detectable ladles
Pelles
For fast and efficient surface sampling use the metal and x ray detectable Ladle.
  • Robust PP construction gives excellent chemical resistance to many products
  • Supplied individually bagged and produced in a cleanroom
  • Full batch traceability
They are ideal for most liquids. The one piece moulded design has been specially constructed to go through standard 50,8 mm bungs frequently found on barrels, drums and kegs. This liquid sampling tool is ideal for liquid sampling.
To ensure the cleanliness of the ladles, they are manufactured and packed in aMore Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR