Imprimer
Sack samplers
Fournisseur: Sampling Systems
1320A-01 1320B-01 1320C-01
SAMP1320A-01EA 207 EUR
SAMP1320A-01 SAMP1320B-01 SAMP1320C-01
Sack samplers
Échantillonneurs Préleveurs de solides
These sack samplers are designed to take fast powder samples through the sides of sacks and bags.
  • Suitable for sampling free flowing powders and granules
  • 316L stainless steel construction
  • Unique range of sample collection methods available
Insert the sampler through the side of the sack. Angle sampler downwards to allow the product to flow into the sample collecting container and seal the hole using an ultra adhesive label.
Informations de commande: Includes a 250 ml sampling bottle.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR