Imprimer

Anti-IgG/IgM (H+L) Goat Polyclonal Antibody (Biotin)

Fournisseur: Rockland Immunochemicals
610-106-115
ROCK610-106-115EA 319 EUR
ROCK610-106-115
Anti-IgG/IgM (H+L) Goat Polyclonal Antibody (Biotin)
Anticorps
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Goat
Isotype:
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR