Imprimer

Anti-CD86 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: GL-1]

Fournisseur: Rockland Immunochemicals
200-502-N90
ROCK200-502-N90EA 797 EUR
ROCK200-502-N90
Anti-CD86 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: GL-1]
Anticorps
Anti-CD86 is useful for immunochemistry, Immunofluorescence, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: GL-1
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR