Imprimer

Anti-ZIC1 Rabbit Polyclonal Antibody

Fournisseur: Rockland Immunochemicals
200-401-159
ROCK200-401-159EA 821 EUR
ROCK200-401-159
Anti-ZIC1 Rabbit Polyclonal Antibody
Anticorps
Primary Rabbit Anti-Zic-1 Reacts with Human, Mouse, Rat

  • Recommended Dilutions: ELISA: 1:10000 - 1:50000; Immunofluorescence: 1:400; Western Blot: 1:5000; Immunochemistry: 1:400;

Type: Primary
Antigen: ZIC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR