Imprimer

Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 15F6.F6.F9]

Fournisseur: Rockland Immunochemicals
200-301-I39S
ROCK200-301-I39SEA 226 EUR
ROCK200-301-I39S
Anti-CALCA Mouse Monoclonal Antibody (Unconjugated) [clone: 15F6.F6.F9]
Anticorps
Anti-Procalcitonin antibody is suitbale for use in ELISA and Western Blot. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:1000-1:20000; Western Blot: 1µg/ml;

Type: Primary
Antigen: CALCA
Clonality: Monoclonal
Clone: 15F6.F6.F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 lambda
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR