Imprimer

Anti-FTO Mouse Monoclonal Antibody (Unconjugated) [clone: 5-2H10]

Fournisseur: Rockland Immunochemicals
200-301-D41
ROCK200-301-D41EA 980 EUR
ROCK200-301-D41
Anti-FTO Mouse Monoclonal Antibody (Unconjugated) [clone: 5-2H10]
Anticorps
Anti-FTO (Mouse) Antibody is suitable for use in Western Blotting and IF. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 58 kDa in size corresponding to FTO Proteins in Western blots of Rat testes lysate.

Recommended Dilutions: Immunofluorescence: 1:100; Western Blot: 1:1000

Type: Primary
Antigen: FTO
Clonality: Monoclonal
Clone: 5-2H10
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG
Reactivity: Mouse, Rat, Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR