Imprimer

Anti-PPI Goat Polyclonal Antibody

Fournisseur: Rockland Immunochemicals
200-1174S
ROCK200-1174SEA 226 EUR
ROCK200-1174S
Anti-PPI Goat Polyclonal Antibody
Anticorps
Anti-Papain antibody is suitable for western blotting to detect a single band of the expected apparent molecular weight and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:3000; Other Dilution: User Optimized

Type: Primary
Antigen: PPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Carica papaya
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR