Imprimer
FAME #17 mix (5 components)
Fournisseur: Restek
35023
REST35023EA 119 EUR
REST35023
FAME #17 mix (5 components)
Étalons Étalons pour l'agroalimentaire
This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the types of oils like linseed, perilla, hempseed and rubberseed oil.
This mix contains five components such as C16:00 methyl palmitate (7%), C18:00 methyl stearate (5%), C18:01 methyl oleate (18%), C18:02 methyl linoleate (36%), C18:03 methyl linolenate (34%).
% represents weight/weight% basis (minimum 50 mg/ampoule).
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR