Imprimer
Gas calibration standards
Fournisseur: Restek
34451 34457 34460
REST34451EA 272 EUR
REST34451 REST34457 REST34460
Gas calibration standards
Étalons Étalons pour l'analyse environnementale
These transportable pure gases and mixtures are meant for purging or calibrating to multi component mixes, which are ideal for peak identification work.
The 14 l container has a CGA 160 connection for more precise integration with analytical systems and 48 l cylinder has a CGA 165 connection and can deliver large volumes of sample.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR