Imprimer
Bulk adsorbents, Florisil® PR
Fournisseur: Restek
26135
REST26135EA 0 EUR
REST26135
Bulk adsorbents, Florisil® PR
Sorbants pour chromatographie Sorbants SPE
Florisil® PR is commonly used to remove polar interferences from pesticide residues. This bulk material is ideal for labs packing their own extraction cartridges for pesticide residue extractions.
  • Pesticide residue grade
Conditionnement: Packaged in glass containers.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR