Imprimer
Make life easier (MLE) capillary tool kit for PerkinElmer GCs
Fournisseur: Restek
22185
REST22185EA 483 EUR
REST22185
Make life easier (MLE) capillary tool kit for PerkinElmer GCs
Sets d'outils
MLE tool kits conveniently provide the tools that make it easier to install and maintain capillary columns.
Information de livraison: Kit includes ¹/₈", ³/₁₆", ¹/₄" nylon brushes, ¹/₄", ³/₈", ³/₁₆" stainless steel wire tube brushes, stainless steel surface brush, 6 stainless steel jet reamers (0,25 to 0,65 mm OD), ¹/₄×⁵/₁₆" open end wrench, ³/₈×⁷/₁₆" open end wrench, rubber-tipped slide-lock tweezers, scoring wafers, inlet liner removal tool, septum puller, mini wool puller/inserter tool, 4-inch tapered needle file, swivel head flashlight, mini hand drill set, 15 cm compact steel ruler,More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR