Imprimer
Disposable particle filtering respirators, P100
Fournisseur: Portwest
P100WHR
PRTWP100WHREA 0 EUR
PRTWP100WHR
Disposable particle filtering respirators, P100
Protection respiratoire Masques à usage unique Demi-masques fitrants contre les particules
This disposable respirator is designed for applications involving dust, mists and fibres. The P100 can be used in concentrations of contaminants up to 4 times the WEL.
  • Soft foam nose seal
  • Elasticated twin straps
  • Adjustable nose bridge
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR