Imprimer
Safety boots, pull-on, Steelite™ WORK rigger FW29
Fournisseur: Portwest
FW29BKR44 FW29BKR45 FW29BKR41 FW29BKR42 FW29BKR43 FW29BKR46 FW29BKR47 FW29BKR48 FW29BKR38 FW29BKR39 FW29BKR40 FW29TAR47 FW29TAR48 FW29TAR40 FW29TAR41 FW29TAR42 FW29TAR43 FW29TAR44 FW29TAR45 FW29TAR46 FW29TAR38 FW29TAR39
PRTWFW29BKR44EA 68 EUR
PRTWFW29BKR44 PRTWFW29BKR45 PRTWFW29BKR41 PRTWFW29BKR42 PRTWFW29BKR43 PRTWFW29BKR46 PRTWFW29BKR47 PRTWFW29BKR48 PRTWFW29BKR38 PRTWFW29BKR39 PRTWFW29BKR40 PRTWFW29TAR47 PRTWFW29TAR48 PRTWFW29TAR40 PRTWFW29TAR41 PRTWFW29TAR42 PRTWFW29TAR43 PRTWFW29TAR44 PRTWFW29TAR45 PRTWFW29TAR46 PRTWFW29TAR38 PRTWFW29TAR39
Safety boots, pull-on, Steelite™ WORK rigger FW29
Chaussures et bottes
Full S3 protection with protective scuff cap, steel toecap and mid sole with side tabs for easy donning.
  • Fur lined for extra warmth and comfort
  • Energy absorbing seat region
  • Protective steel toecap
  • Heat resistant outsole: 200 °C
  • Upper material: Buffalo leather
  • Anti-static footwear
Slip resistant outsole to prevent slips and trips on ceramic and steel surfaces.
Certifications: EN ISO 20345:2011
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR