Imprimer
Fall protection harnesses, 1 point
Fournisseur: Portwest
FP11RER
PRTWFP11REREA 26.2 EUR
PRTWFP11RER
Fall protection harnesses, 1 point
Protections anti-chutes Harnais de sécurité
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
  • Dorsal D-ring
  • Chest and leg adjustment points
  • Flat bar buckles
  • Inspection cover for protection of product label
  • Retail box which aids presentation for retail sales
  • Made from polyester webbing, alloy steel, heat treated
Ideal for fall arrest or work restraint systems, our entry level full body harness is light weight and easy to use, allowing a secure fit.
Certifications: EN 361
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR