Imprimer
Gloves, nitrile, Flexo Grip, A310
Fournisseur: Portwest
A310GRWL A310GRWXL A310R8RXL A310R8RXXL A310GRWXXL A310GRWM A310GRWS A310R8RL A310R8RM A310R8RS
PRTWA310GRWLEA 12.1 EUR
PRTWA310GRWL PRTWA310GRWXL PRTWA310R8RXL PRTWA310R8RXXL PRTWA310GRWXXL PRTWA310GRWM PRTWA310GRWS PRTWA310R8RL PRTWA310R8RM PRTWA310R8RS
Gloves, nitrile, Flexo Grip, A310
Gants
The nitrile (pylon) coating on the Flexo Grip glove gives enhanced abrasion resistance. Ideal for auto repair, construction and other sectors.
  • Maximum EN level 4 abrasion
  • Palm dipped to increase dexterity and ventilation
  • 13 gauge liner for a perfect fit
  • 100% Breathable seamless liner
  • Low linting construction for minimal contamination
  • Retail bag which aids presentation for retail sales
ANSI Abrasion level 2 and cut level A1.
Certifications: AS/NZS 2161 .2, AS/NZS 2161 .3, EN 388:2016 - (3.1.2.1.X), ANSI/ISEA 105 - 2011 Abrasion Level (2), EN 420
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR