Imprimer
Peanut butter, NIST-2387 standard
Fournisseur: LGC Standards PROMOCHEM
NIST-2387
PROONIST-2387EA 2470 EUR
PROONIST-2387
Peanut butter, NIST-2387 standard
Étalons Étalons pour l'agroalimentaire
This reference material intended to use in validating methods for determining fat, fatty acids, trace elements and tocopherol.
Certifications: Traceable to NIST and SRM.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR