Imprimer
Cleaning brush
Fournisseur: PFENNIG REINIGUNGSTECHNIK
3590500
PPSA3590500EA 35.6 EUR
PPSA3590500
Cleaning brush
Brosses
Handy brush with long handle for hygienic sensitive areas.
  • Material: PP (body), PBT (stock)
  • Stock height 35 mm
  • Unslit fibre ends
  • Hard bristle material
Colour: White
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR