Imprimer

S-Methyl methanethiosulphonate (MMTS), Pierce™

Fournisseur: Thermo Fisher Scientific

Synonyms: Methanethiosulphonic Acid S-Methyl Ester, Methyl methanethiosulphonate

23011
PIER23011EA 152 EUR
PIER23011
S-Methyl methanethiosulphonate (MMTS), Pierce™
S-Methyl methanethiosulphonate (MMTS)

Pierce™ MMTS (methyl methanethiosulfonate) is a small compound that reversibly blocks cysteines and other sulfhydryl groups, enabling the study of enzyme activation and other protein functions.


  • Converts sulfhydryl groups on cysteine side chains into -S-S-CH3
  • Reaction is reversible with DTT or TCEP, restoring the free sulfhydryl
  • Used to modify thiol groups in creatine kinase

Formule: C₂H₆O₂S₂
MW: 126.19 g/mol
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00007565
Numéro CAS: 2949-92-0

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR