Imprimer
Nitrogen generator, Infinity 1031
Fournisseur: PEAK SCIENTIFIC
13-1270
PEAK13-1270EA 14360 EUR
PEAK13-1270
Nitrogen generator, Infinity 1031
Générateurs de gaz Générateurs d'azote
Membrane technology is at the heart of the Infinity 1031, generating laboratory-grade nitrogen on-site with an existing air supply. With the ability to perform at its maximum 24 hours a day, if required, and minimum maintenance requirements, the Infinity 1031 is a durable, hassle-free solution for Sciex LC-MS instruments.
  • 24/7 operation at optimum performance if required
  • Gas is supplied on demand so generator works to your schedule
  • Few moving parts means little maintenance required and ensures long life of the generator
  • Minimal set-up required
  • Completely
More Product Information
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR