Imprimer
Quanti-Cult Plus™ Bacillus subtilis ATCC™ 6633™
Fournisseur: THERMO FISHER DIAGNOSTICS
R4711221
OXOIR4711221EA 783 EUR
OXOIR4711221
Quanti-Cult Plus™ Bacillus subtilis ATCC™ 6633™
Tests microbiologiques Tests sur les micro-organismes
Quanti-Cult Plus™ Bacillus subtilis ATCC™ 6633™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.1 ml inoculum
  • 10 tests/reconstituted vial; 100 tests/kit
  • Ready for rehydration
  • For use in quality control procedures
Conditionnement: Includes 10 preserved organism vials and 10 rehydration vials.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR