Imprimer
Culti-Loops™ Microbacterium liquefaciens ATCC™ BAA-1819™
Fournisseur: THERMO FISHER DIAGNOSTICS
R4609417
OXOIR4609417EA 178 EUR
OXOIR4609417
Culti-Loops™ Microbacterium liquefaciens ATCC™ BAA-1819™
Tests microbiologiques Tests sur les micro-organismes
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Conditionnement: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR