Imprimer
Culti-Loops™ Staphylococcus epidermidis ATCC™ 49134™
Fournisseur: THERMO FISHER DIAGNOSTICS
R4607017
OXOIR4607017EA 178 EUR
OXOIR4607017
Culti-Loops™ Staphylococcus epidermidis ATCC™ 49134™
Tests microbiologiques Tests sur les micro-organismes
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Conditionnement: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Commander maintenant
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR