Imprimer
Culti-Loops™ Streptococcus criceti ATCC™ 19642™
Fournisseur: THERMO FISHER DIAGNOSTICS
R4606902
OXOIR4606902EA 178 EUR
OXOIR4606902
Culti-Loops™ Streptococcus criceti ATCC™ 19642™
Tests microbiologiques Tests sur les micro-organismes
Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium or streaked directly onto an appropriate media.
Conditionnement: Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
Plus About VWR