Imprimer
PCR expansion
Fournisseur: Oxford Nanopore Technologies
EXP-PCA001
OXNTEXP-PCA001EA 420 EUR
OXNTEXP-PCA001
PCR expansion
Réactifs pour acides nucléiques Sets de réactifs pour séquençage de nouvelle génération
PCR Primers (PRM) and PCR Adapters (PCA).
  • Additional PCR primers and adapters for use with Ligation Sequencing Kits
This kit contains PCR Primers and PCR Adapters, sufficient for at least 24 reactions, and is compatible for use with the Ligation Sequencing Kits (OXNTSQK-LSK110 and OXNTSQK-LSK109).
Information de livraison: Flow cells and kits are shipped together at 2 to 8 °C. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be three months from receipt by the customer.
Attention: Oxford Nanopore Technologies products are currently for research use only.
Commander maintenant
Please note that Oxford Nanopore Technologies requires the acceptance of their Terms & Conditions prior to product order placement. If you select product(s) to order, the Terms and Conditions will be displayed in the shopping cart for your review and acceptance.
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Plus About VWR